missing translation for 'onlineSavingsMsg'
Learn More

Amaranth, MP Biomedicals

Product Code. 11411106 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g

Product Code. 11411106

Brand: MP Biomedicals 0520245380

Please to purchase this item. Need a web account? Register with us today!

Specifications

CAS 915-67-3
Molecular Formula C20H11N2Na3O10S3
MDL Number MFCD00004076
Quantity 100 g
Synonym amaranth, azo rubin, amaranth, analytical standard, amaranth azorubin s, amaranth, dye content 85-95 %, trisodium 3-hydroxy-4-e-2-4-sulfonatonaphthalen-1-yl diazen-1-yl naphthalene-2,7-disulfonate, trisodium 3-oxo-4-4-sulfonatonaphthalen-1-yl hydrazinylidene naphthalene-2,7-disulfonate
InChI Key VOBHRQFELWTZFS-DXVNCNPQSA-K
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=C1)C=C(C(=O)\C2=N\NC1=C2C=CC=CC2=C(C=C1)S([O-])(=O)=O)S([O-])(=O)=O
IUPAC Name trisodium (4E)-3-oxo-4-[2-(4-sulfonatonaphthalen-1-yl)hydrazin-1-ylidene]-3,4-dihydronaphthalene-2,7-disulfonate
Molecular Weight (g/mol) 604.46
PubChem CID 6364527
Formula Weight 604.49
Chemical Name or Material Amaranth
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.