missing translation for 'onlineSavingsMsg'
Learn More

Amantadine hydrochloride, MedChemExpress

Product Code. 16444357
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
This item is not returnable. View return policy

Product Code. 16444357

Brand: Medchem Express HYB0402A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Amantadine (1-Adamantanamine) hydrochloride is an orally avtive and potent antiviral agent with activity against influenza A viruses.

Amantadine hydrochloride inhibits several ion channels such as NMDA and M2, and also inhibits Coronavirus ion channels. Amantadine hydrochloride also has anti-orthopoxvirus and anticancer activity. Amantadine hydrochloride can be used for Parkinson's disease, postoperative cognitive dysfunction (POCD) and COVID-19 research.

Specifications

Chemical Name or Material Amantadine hydrochloride
Physical Form Solid
CAS 665-66-7
Color White
Quantity 50 g
Purity Grade Notes Research
Molecular Formula C10H18ClN
For Use With (Application) COVID-19-anti-virus
Synonym 1-Adamantanamine hydrochloride 1-Adamantylamine hydrochloride 1-Aminoadamantane hydrochloride
Solubility Information DMSO : 100 mg/mL (532.74 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (266.37 mM)
SMILES NC1(C2)C[C@@H]3C[C@@H](C[C@H]2C3)C1.Cl
Molecular Weight (g/mol) 187.71
Formula Weight 187.71
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.