missing translation for 'onlineSavingsMsg'
Learn More

AM966, MedChemExpress

Product Code. 16476706
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476706 100 mg 100mg
16456418 10 mg 10mg
16439007 5 mg 5mg
16422527 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476706 Supplier Medchem Express Supplier No. HY15277
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AM966 is a high affinity, selective, oral LPA1-antagonist, inhibits LPA-stimulated intracellular calcium release (IC50=17 nM).

AM966 is a high affinity, selective, oral LPA1-antagonist, inhibits LPA-stimulated intracellular calcium release (IC50=17 nM).

Specifications

Chemical Name or Material AM966
CAS 1228690-19-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H23ClN2O5
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (203.70 mM; Need ultrasonic)
SMILES O=C(O)CC1=CC=C(C2=CC=C(C3=C(NC(O[C@@H](C4=CC=CC=C4Cl)C)=O)C(C)=NO3)C=C2)C=C1
Molecular Weight (g/mol) 490.93
Formula Weight 490.93
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.