missing translation for 'onlineSavingsMsg'
Learn More

AM679, MedChemExpress

Product Code. 16488677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488677 5 mg 5mg
16456078 10 mg 10mg
16442217 50 mg 50mg
16436476 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488677 Supplier Medchem Express Supplier No. HY14460

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AM679 is a potent, selective 5-lipoxygenase-activating protein (FLAP) inhibitor with an IC50 of 2 nM in a human FLAP membrane binding assay. AM679 markedly reduces the respiratory syncytial virus-driven ocular pathology as well as the synthesis of cysteinyl leukotrienes (CysLTs) in the eye.

AM679 is a potent, selective 5-lipoxygenase-activating protein (FLAP) inhibitor with an IC50 of 2 nM in a human FLAP membrane binding assay. AM679 markedly reduces the respiratory syncytial virus-driven ocular pathology as well as the synthesis of cysteinyl leukotrienes (CysLTs) in the eye.

Specifications

Chemical Name or Material AM679
CAS 1206880-66-1
Color pale pink
Purity Grade Notes Research
Molecular Formula C40H44N4O5S
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (144.33 mM)
SMILES O=C(C)N1C2=CC=CC=C2C[C@H]1COC3=CC=C(N(CC4=CC=C(C5=NC=C(OC)C=N5)C=C4)C(CC(C)(C(O)=O)C)=C6SC(C)(C)C)C6=C3
Molecular Weight (g/mol) 692.87
Formula Weight 692.87
Percent Purity 99.6%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.