missing translation for 'onlineSavingsMsg'
Learn More

AM580, MedChemExpress

Product Code. 16480347
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480347 50 mg 50mg
16456707 5 mg 5mg
16433998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16480347 Supplier Medchem Express Supplier No. HY10475
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AM580 is a selective RARα agonist with IC50 and EC50 of 8 nM and 0.36 nM, respectively.

AM580 is a selective RARα agonist with IC50 and EC50 of 8 nM and 0.36 nM, respectively.

Specifications

Chemical Name or Material AM580
CAS 102121-60-8
Color White
Purity Grade Notes Research
Molecular Formula C22H25NO3
Quantity 50 mg
Synonym CD336 NSC608001 Ro 40-6055
Solubility Information DMSO : ≥ 45 mg/mL (128.04 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC(CC1)(C)C2=C(C=CC(C(NC3=CC=C(C(O)=O)C=C3)=O)=C2)C1(C)C
Molecular Weight (g/mol) 351.44
Formula Weight 351.44
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.