missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies AM580
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- AM580 is a retinoic acid receptor (RAR) agonist that is selective for RARα (EC50 = 0.36 nM) compared to RARβ (EC50 = 24.6 nM) and RARγ (EC50 = 27.9 nM; Bernard et al.) It is a derivative of retinoic acid (RA), however it demonstrates greater specific binding to RARα compared to RA, which exhibits little selectivity across RARα, β, or γ (Gianní et al.; Bernard et al.; Kim et al; Rochette-Egly & Germain)
- Promotes reprogramming of somatic cells to induced pluripotent stem cells (Wang et al.)
- Induces differentiation of human induced pluripotent stem cells into intermediate mesoderm, in combination with the GSK3β inhibitor CHIR99021 (Araoka et al.)
- Inhibits tumor cell proliferation and survival signaling pathways, and induces apoptosis, leading to inhibition of mouse mammary tumor virus (MMTV)-neu- and MMTV-wnt1-induced mammary gland hyperplasia (Lu et al.)
- Inhibits tumor growth in MMTV-Myc mice (Bosch et al.)
- Inhibits endometrial cancer cell proliferation (Cheng et al.)
- Induces differentiation in acute promyelocytic leukemia cells (Gianní et al.)
Specifications
Specifications
| CAS | 102121-60-8 |
| Synonym | CD336; NSC 608001; Ro 40-6055 |
| Quantity | 10 mg |
| Molecular Formula | C22H25NO3 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction