missing translation for 'onlineSavingsMsg'
Learn More

AM251, MedChemExpress

Product Code. 16406528
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16406528

Brand: Medchem Express HY15443

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AM251 is a selective cannabinoid 1 (CB1) receptor antagonist with an IC50 of 8 nM. AM251 also acts as a potent GPR55 agonist with an EC50 of 39 nM.

AM251 is a selective cannabinoid 1 (CB1) receptor antagonist with an IC50 of 8 nM. AM251 also acts as a potent GPR55 agonist with an EC50 of 39 nM.

Specifications

Chemical Name or Material AM251
CAS 183232-66-8
Color White
Purity Grade Notes Research
Molecular Formula C22H21Cl2IN4O
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (45.03 mM; Need ultrasonic)
SMILES O=C(C1=NN(C2=CC=C(Cl)C=C2Cl)C(C3=CC=C(I)C=C3)=C1C)NN4CCCCC4
Molecular Weight (g/mol) 555.24
Formula Weight 555.24
Percent Purity 98.82%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.