missing translation for 'onlineSavingsMsg'
Learn More

AM251, MedChemExpress

Product Code. 16469107
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469107 5 mg 5mg
16486766 100 mg 100mg
16462617 50 mg 50mg
16406528 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16469107 Supplier Medchem Express Supplier No. HY15443
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AM251 is a selective cannabinoid 1 (CB1) receptor antagonist with an IC50 of 8 nM. AM251 also acts as a potent GPR55 agonist with an EC50 of 39 nM.

AM251 is a selective cannabinoid 1 (CB1) receptor antagonist with an IC50 of 8 nM. AM251 also acts as a potent GPR55 agonist with an EC50 of 39 nM.

Specifications

Chemical Name or Material AM251
CAS 183232-66-8
Color White
Purity Grade Notes Research
Molecular Formula C22H21Cl2IN4O
Quantity 5 mg
Solubility Information DMSO : 25 mg/mL (45.03 mM; Need ultrasonic)
SMILES O=C(C1=NN(C2=CC=C(Cl)C=C2Cl)C(C3=CC=C(I)C=C3)=C1C)NN4CCCCC4
Molecular Weight (g/mol) 555.24
Formula Weight 555.24
Percent Purity 98.82%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.