missing translation for 'onlineSavingsMsg'
Learn More

AM211, MedChemExpress

Product Code. 16421727
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16421727

Brand: Medchem Express HY13213

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AM211 is a potent, selective and orally bioavailable prostaglandin D2 (PGD2) receptor type 2 (DP2) antagonist, with IC50s of 4.9 nM, 7.8 nM, 4.9 nM, 10.4 nM for human, mouse, guinea pig, and rat DP2, respectively.

AM211 is a potent, selective and orally bioavailable prostaglandin D2 (PGD2) receptor type 2 (DP2) antagonist, with IC50s of 4.9 nM, 7.8 nM, 4.9 nM, 10.4 nM for human, mouse, guinea pig, and rat DP2, respectively.

Specifications

Chemical Name or Material AM211
CAS 1175526-27-8
Color White
Purity Grade Notes Research
Molecular Formula C27H27F3N2O4
Quantity 50 mg
Synonym AM211 free acid
Solubility Information DMSO : ≥ 125 mg/mL (249.75 mM)
SMILES O=C(O)CC1=CC(C2=CC=C(C(F)(F)F)C=C2CN(CC)C(NCC3=CC=CC=C3)=O)=C(OC)C=C1
Molecular Weight (g/mol) 500.51
Formula Weight 500.51
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.