missing translation for 'onlineSavingsMsg'
Learn More

AM-4668, MedChemExpress

Product Code. 16417977
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16417977 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16417977 Supplier Medchem Express Supplier No. HY12585
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AM-4668 is a GPR40 agonist for type 2 diabetes. EC50s of 3.6 nM and 36 nM for GPR40 in A9 cells (GPR40 IP3 assay) and CHO cells (GPR40 aequorin assay), respectively.

AM-4668 is a GPR40 agonist for type 2 diabetes. EC50s of 3.6 nM and 36 nM for GPR40 in A9 cells (GPR40 IP3 assay) and CHO cells (GPR40 aequorin assay), respectively.

Specifications

Chemical Name or Material AM-4668
CAS 1011531-27-3
Color White
Purity Grade Notes Research
Molecular Formula C24H19F3N2O4S
Quantity 5 mg
SMILES OC(C[C@H](C1=NOC=C1)C(C=C2)=CC=C2OCC(S3)=C(C)N=C3C4=CC=C(C(F)(F)F)C=C4)=O
Molecular Weight (g/mol) 488.48
Formula Weight 488.48
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.