missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALY Recombinant Rabbit Monoclonal Antibody (23GB5660), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | ALY |
|---|---|
| Clone | 23GB5660 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089226
![]() |
Invitrogen™
MA552888 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089171
|
Invitrogen™
MA552889 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALY Recombinant Monoclonal antibody specifically detects ALY in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| ALY | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O08583, Q86V81 | |
| 10189, 21681, 690585 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5660 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ally of AML-1 and LEF-1; ALY; ALY/REF; Aly/REF export factor; Aly/REF export factor L homeolog; aly/REF export factor-A; alyref; alyref.L; alyref-a; BEF; bZIP enhancing factor; bZIP-enhancing factor BEF; hypothetical protein LOC537706; REF; Ref1; REF1-I; Refbp1; RNA and export factor binding protein 1; RNA and export factor-binding protein 1; Tcra enhancer-binding factor interacting protein 1; THO complex 4; THO complex subunit 4; THO complex subunit 4-A; tho4; Tho4-A; THOC4; thoc4-a; transcriptional coactivator Aly/REF; XELAEV_18043750mg; zgc:171753 | |
| A synthesized peptide derived from human Aly (201-257AA). | |
| Primary | |
| -20°C | |
| ALYREF |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title