missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Aly Polyclonal specifically detects Aly in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Aly |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | Ally of AML-1 and LEF-1, ALY/REF, ALYBEFbZIP enhancing factor, bZIP-enhancing factor BEF, REF, THO complex 4, THO complex subunit 4, tho4, Transcriptional coactivator Aly/REF |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N terminal region of mouse REFBP2 (NP_062357.3). Peptide sequence KMDMSLDDIIKLNRNQRRVNRGGGPRRNRPAIARGGRNRPAPYSRPKPLP |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?