missing translation for 'onlineSavingsMsg'
Learn More

ALY Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA582697

Product Code. 16379654

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EFGTLKKAAV HYDRSGRSLG TADVHFERKA DALKAMKQYN GVPLDGRPMN IQLVTSQIDA QRRPAQSVNR GGMTRNRGAG GFGGGGGTRR GTRGGARGRG RGAGRNSKQQ LSAEELDAQL DAYNARMD

The protein encoded by this gene is a heat stable, nuclear protein and functions as a molecular chaperone. It is thought to regulate dimerization, DNA binding, and transcriptional activity of basic region-leucine zipper (bZIP) proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ALY
Polyclonal
Unconjugated
ALYREF
ALY, ALY/REF, BEF, REF, THOC4
Rabbit
Affinity Chromatography
RUO
10189
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.10 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q86V81
ALYREF
Recombit protein corresponding to Human ALYREF
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.