missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALY Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542286
This item is not returnable.
View return policy
Description
Peptide sequence: GGGPIRNRPA IARGAAGGGG RNRPAPYSRP KQLPDKWQHD LFDSGFGGGA Sequence homology: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.
The protein encoded by this gene is a heat stable, nuclear protein and functions as a molecular chaperone. It is thought to regulate dimerization, DNA binding, and transcriptional activity of basic region-leucine zipper (bZIP) proteins.
Specifications
| ALY | |
| Polyclonal | |
| Unconjugated | |
| ALYREF | |
| ally of AML-1 and LEF-1; ALY; ALY/REF; Aly/REF export factor; Aly/REF export factor L homeolog; aly/REF export factor-A; alyref; alyref.L; alyref-a; BEF; bZIP enhancing factor; bZIP-enhancing factor BEF; hypothetical protein LOC537706; REF; Ref1; REF1-I; Refbp1; RNA and export factor binding protein 1; RNA and export factor-binding protein 1; Tcra enhancer-binding factor interacting protein 1; THO complex 4; THO complex subunit 4; THO complex subunit 4-A; tho4; Tho4-A; THOC4; thoc4-a; transcriptional coactivator Aly/REF; XELAEV_18043750mg; zgc:171753 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10189 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q86V81 | |
| ALYREF | |
| Synthetic peptide directed towards the N-terminal of human THOC4 (aa 51-100). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction