Learn More
Invitrogen™ ALY Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596489
Description
Positive Samples: HeLa, A-549, B cells, SKOV3, HL-60, Mouse testis, Mouse spleen, Mouse thymus, Rat brain; Cellular Location: Cytoplasm, Nucleus, Nucleus speckle Immunogen sequence: MPDSAPAMAD KMDMSLDDII KLNRSQRGGR GGGRGRGRAG SQGGRGGGAQ AAARVNRGGG PIRNRPAIAR GAAGGGGRNR PAPYSRPKQL PDKWQHDLFD.
The protein encoded by this gene is a heat stable, nuclear protein and functions as a molecular chaperone. It is thought to regulate dimerization, DNA binding, and transcriptional activity of basic region-leucine zipper (bZIP) proteins.
Specifications
| ALY | |
| Polyclonal | |
| Unconjugated | |
| ALYREF | |
| ally of AML-1 and LEF-1; ALY; ALY/REF; Aly/REF export factor; Aly/REF export factor L homeolog; aly/REF export factor-A; alyref; alyref.L; alyref-a; BEF; bZIP enhancing factor; bZIP-enhancing factor BEF; hypothetical protein LOC537706; REF; Ref1; REF1-I; Refbp1; RNA and export factor binding protein 1; RNA and export factor-binding protein 1; Tcra enhancer-binding factor interacting protein 1; THO complex 4; THO complex subunit 4; THO complex subunit 4-A; tho4; Tho4-A; THOC4; thoc4-a; transcriptional coactivator Aly/REF; XELAEV_18043750mg; zgc:171753 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10189, 21681, 690585 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.48 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O08583, Q86V81 | |
| ALYREF | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human THOC4/THOC4/ALYREF (NP_0057733). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.