Learn More
Invitrogen™ ALX4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113569
Description
Immunogen sequence: GQTHMGSLFG AASLSPGLNG YELNGEPDRK TSSIAALRMK AKEHSAAISW For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 100%.
This gene encodes a paired-like homeodomain transcription factor expressed in the mesenchyme of developing bones, limbs, hair, teeth, and mammary tissue. Mutations in this gene cause parietal foramina 2 (PFM2); an autosomal dominant disease characterized by deficient ossification of the parietal bones. Mutations in this gene also cause a form of frontonasal dysplasia with alopecia and hypogonadism; suggesting a role for this gene in craniofacial development, mesenchymal-epithelial communication, and hair follicle development. Deletion of a segment of chromosome 11 containing this gene, del(11)(p11p12), causes Potocki-Shaffer syndrome (PSS); a syndrome characterized by craniofacial anomalies, mental retardation, multiple exostoses, and genital abnormalities in males. In mouse, this gene has been shown to use dual translation initiation sites located 16 codons apart.
Specifications
| ALX4 | |
| Polyclonal | |
| Unconjugated | |
| ALX4 | |
| ALX homeobox 4; ALX4; ALX-4; aristaless 4; Aristaless-like 4; aristaless-like homeobox 4; CRS5; FND2; Homeobox protein aristaless-like 4; homeodomain transcription factor ALX4; KIAA1788; lst; Strong's luxoid | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 60529 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H161 | |
| ALX4 | |
| Synthetic peptide directed towards the middle region of human ALX4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.