missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALX3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536138
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 36 kDa. Purity is >95% by SDS-PAGE.
Aristaless-related genes are a group of paired-related homeobox genes which play a role in regulating vertebrate embryogenesis. The homeodomain transciption factor aristaless-like 3 (ALX3) is expressed in mouse embryos from 8 days of gestation, predominantly in neural crest-derived mesenchyme and in lateral plate mesoderm. Expression analysis of human and mouse tissue reveals predominant ALX3 expression in brain tissue. The Alx3 gene maps to chromosome 1p23-p13 and encodes a 343 amino acid protein. Preferential methylation of Alx3 occurs in advanced-stage neuroblastoma and may repress ALX3 expression. Treatment with the methylation inhibitor 5-aza-2'-deoxycytidine restores ALX3 expression. Alx3 (-) mice lack a phenotype distinct from wild-type mice, however Alx3/Alx4 double mutants demonstrate severe craniofacial abnormalities not present in Alx4 single mutants. Specifically, Alx3/Alx4 double mutant newborn mice have cleft nasal regions in addition to malformation of other neural crest-derived skull structures.
Specifications
| ALX3 | |
| Polyclonal | |
| Unconjugated | |
| ALX3 | |
| ALX homeobox 3; ALX3; aristaless 3; aristaless-like homeobox 3; FND; FND1; frontonasal dysplasia; homeobox protein aristaless-like 3; Proline-rich transcription factor ALX3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11694, 257, 365900 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O70137, O95076 | |
| ALX3 | |
| Synthetic peptide corresponding to amino acids 171-220 of human ALX3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction