missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALX3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80285
This item is not returnable.
View return policy
Description
ALX3 Polyclonal specifically detects ALX3 in Mouse samples. It is validated for Western Blot.
Specifications
| ALX3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ALX homeobox 3, aristaless-like homeobox 3, homeobox protein aristaless-like 3, MGC138212, MGC141988, Proline-rich transcription factor ALX3 | |
| Rabbit | |
| 37 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Mouse: 100%; Bovine: 92%; Rat: 92%; Human: 78%. | |
| Human, Mouse, Rat, Bovine, Guinea Pig | |
| Purified |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O70137 | |
| ALX3 | |
| Synthetic peptide directed towards the N terminal of mouse ALX3 (NP_031467). Peptide sequence MDPERCAPFSVGPAAGPYAAAGDEAPGPQGTPDAAPHLHPAPPRGPRLSR. | |
| Protein A purified | |
| RUO | |
| 257 | |
| Centrifuge vial prior to reconstitution. Add 100μL distilled water to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction