missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALX1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551608
This item is not returnable.
View return policy
Description
Immunogen sequence: DNESFYSKAS AGKCVQAFGP LPRAEHHVRL ERTSPCQDSS VNYGITKVEG QPLHTELNRA MDNCNSLRMS PVKGMQEKGE LDELGDKCDS NVSSSKKRRH RTTFTSLQLE ELEKVFQKTH YP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
Transcriptional activator that acts at a palindromic recognition sequence to enhance the activity of the SV40 and TK promoters. Functions as a repressor with the prolactin promoter in vivo. May play a role in chondrocyte differentiation and may also influence cervix development.
Specifications
| ALX1 | |
| Polyclonal | |
| Unconjugated | |
| ALX1 | |
| ALX homeobox protein 1, Cartilage homeoprotein 1, CART-1, ALX1, CART1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8092 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15699 | |
| ALX1 | |
| Recombit protein corresponding to Human ALX1 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction