missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alvimopan dihydrate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Alvimopan dihydrate (ADL 8-2698 dihydrate) is a potent, selective, orally active and reversible μ-opioid receptor antagonist, with an IC50 of 1.7 nM. Alvimopan dihydrate has selectivity for μ-opioid receptor (Ki=0.47 nM) over κ- and δ-opioid receptors (Kis=100, 12 nM, respectively). Alvimopan dihydrate can be used for the research of postoperative ileus.
Specifications
Specifications
| Chemical Name or Material | Alvimopan dihydrate |
| CAS | 170098-38-1 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H36N2O6 |
| Quantity | 10 mg |
| Synonym | ADL 8-2698 dihydrate LY 246736 dihydrate |
| Solubility Information | DMSO : 33.33 mg/mL (72.37 mM; Need ultrasonic) |
| SMILES | OC1=CC=CC([C@@]2(C)[C@@H](C)CN(C[C@H](CC3=CC=CC=C3)C(NCC(O)=O)=O)CC2)=C1.O.O |
| Molecular Weight (g/mol) | 460.56 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction