missing translation for 'onlineSavingsMsg'
Learn More

Alvespimycin hydrochloride, MedChemExpress

Product Code. 16483276
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
This item is not returnable. View return policy

Product Code. 16483276

Brand: Medchem Express HY12024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Alvespimycin hydrochloride (17-DMAG hydrochloride; KOS-1022; BMS 826476) is a potent inhibitor of Hsp90, binding to Hsp90 with EC50 of 62±29 nM.

Alvespimycin hydrochloride (17-DMAG hydrochloride

Specifications

Chemical Name or Material Alvespimycin hydrochloride
Physical Form Powder
CAS 467214-21-7
Color Purple
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C32H49ClN4O8
For Use With (Application) Cancer-programmed cell death
Synonym 17-DMAG hydrochloride KOS-1022 BMS 826476
Solubility Information DMSO : ≥ 50 mg/mL (76.55 mM) ∣H2O : 4.76 mg/mL (7.29 mM; ultrasonic and warming and heat to 60°C)
SMILES C/C1=C\C=C/[C@@H]([C@H](/C(C)=C/[C@@H]([C@H]([C@H](C[C@@H](CC(C(C(NC1=O)=CC2=O)=O)=C2NCCN(C)C)C)OC)O[H])C)OC(N)=O)OC.Cl
Molecular Weight (g/mol) 653.21
Formula Weight 653.21
Percent Purity 95.61%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.