missing translation for 'onlineSavingsMsg'
Learn More

Aluminum wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.999% (metals basis)

Product Code. 11376837
Change view
Click to view available options
Quantity:
100 m
25 m
300 m
Unit Size:
100m
25m
300m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376837 300 m 300m
11366837 25 m 25m
11386837 100 m 100m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11376837 Supplier Thermo Scientific Alfa Aesar Supplier No. 010747.H8
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Form: approx. 2.1g/m; Temper: soft | CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifications

Color Silver
Appearance Silver-colored wire
Quantity 300 m
Odor Odorless
Assay Percent Range 99.999% (Metals basis)
Form Wire
IUPAC Name aluminium
Chemical Name or Material Aluminum, Annealed

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.