missing translation for 'onlineSavingsMsg'
Learn More

Aluminum wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11356837
Click to view available options
Quantity:
5 m
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7429-90-5
Density ≈2.1g/m
Molecular Formula Al
MDL Number MFCD00134029
Quantity 5 m
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES [Al]
IUPAC Name aluminum
Molecular Weight (g/mol) 26.98
PubChem CID 5359268
ChEBI CHEBI:33629
Percent Purity 99.999%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Aluminum wire, 1.0mm (0.04 in.) dia, annealed, Puratronic™
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'