missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Aluminum Weighing Scoop

Weigh and measure your chemicals with Bochem™ Aluminum Weighing Scoop for total accuracy.

Brand:  Bochem™ 12810

Code : A0

Additional Details : Weight : 0.22500kg

 View more versions of this product

Product Code. 10376001

  • £5.05 / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

  • Aluminum
Specifications

Specifications

Conical
Conical
70 mm
Stainless Steel
Weighing Scoop
32 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers