missing translation for 'onlineSavingsMsg'
Learn More

Aluminum trifluoromethanesulfonate, 99%

Product Code. 11425686
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425686 25 g 25g
11415686 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425686 Supplier Thermo Scientific Alfa Aesar Supplier No. B20785.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 74974-61-1 | C3AlF9O9S3 | 474.171 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74974-61-1
Molecular Formula C3AlF9O9S3
Molecular Weight (g/mol) 474.171
MDL Number MFCD00143596
InChI Key FKOASGGZYSYPBI-UHFFFAOYSA-K
Synonym aluminum trifluoromethanesulfonate, aluminum triflate, aluminum triflate, aluminum tris trifluoromethanesulfonate, aluminum trifluoromethanesulphonate, aluminum 3+ tritriflate, aluminum trifluoromethanesulfonate, aluminum iii triflate, aluminum iii triflate, aluminum tris fluoranyl methanesulfonate
PubChem CID 2737634
IUPAC Name aluminum;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Al+3]

Specifications

Melting Point ∼300°C
Linear Formula Al(CF3SO3)3
Quantity 25 g
UN Number UN3261
Sensitivity Hygroscopic
Formula Weight 474.2
Percent Purity 99%
Chemical Name or Material Aluminum trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.