missing translation for 'onlineSavingsMsg'
Learn More

Aluminum titanium oxide, 99.5% (metals basis)

Product Code. 11312438
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11312438

Brand: Thermo Scientific Alfa Aesar 014484.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12004-39-6 | Al2O5Ti | 181.825 g/mol

Aluminum titanium oxide is used in refractory industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12004-39-6
Molecular Formula Al2O5Ti
Molecular Weight (g/mol) 181.825
MDL Number MFCD00064749
InChI Key CNRZQDQNVUKEJG-UHFFFAOYSA-N
Synonym aluminum titanate, powder, oxo-bis oxoalumanyloxy titanium, trioxo-2,4-dioxa-1,5-dialumina-3-titanapentane, aluminum titanate, nanopowder, <25 nm particle size bet trace metals basis
PubChem CID 16213786
IUPAC Name oxo-bis(oxoalumanyloxy)titanium
SMILES O=[Al]O[Ti](=O)O[Al]=O

Specifications

Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 181.86
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Aluminum titanium oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.