missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Thinfoil, 2.0 micron (0.00008 in.) thick, 99.1% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7051189
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
[Al]
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
[Al]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 x 50 mm
IUPAC Name aluminium
Percent Purity 99.1%
Odor Odorless
Particle Size 2.0 Micron (0.00008 in.) thick
Weight ≈0.013g/50x50mm
Form Foil
Assay (metals basis)
Chemical Name or Material Aluminum Thinfoil, Not certified pinhole free

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'