missing translation for 'onlineSavingsMsg'
Learn More

Aluminum tert-Butoxide, TRC

Product Code. 30308898
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30308898 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30308898 Supplier Toronto Research Chemicals Supplier No. TRCA5758905G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminum tert-Butoxide
CAS 556-91-2
Molecular Formula C4H10O.1/3Al
Quantity 5 g
Synonym Aluminum tert-butoxide (6CI,7CI), tert-Butyl Alcohol Aluminum Salt, Aluminum t-Butoxide, Aluminum tert-Butanolate, Aluminum tert-Butylate, Aluminum tri-tert-Butoxide, Aluminum tris(tert-Butoxide), Aluminum tris(tert-Butylate), Tri-tert-Butoxyaluminum, Tris(2-methyl-2-propanolato)aluminum
InChI Formula InChI=1S/3C4H9O.Al/c3*1-4(2,3)5;/h3*1-3H3;/q3*-1;+3
SMILES CC(C)(C)O[Al](OC(C)(C)C)OC(C)(C)C
IUPAC Name tris[(2-methylpropan-2-yl)oxy]alumane
Molecular Weight (g/mol) 83.11
Formula Weight 246.1776
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.