missing translation for 'onlineSavingsMsg'
Learn More

Aluminum tert-butoxide, 97%

Product Code. 11413850
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11413850

Brand: Thermo Scientific Alfa Aesar 089891.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 556-91-2 | C12H27AlO3-3 | 246.327 g/mol

Aluminum tert-butoxide is utilized as a reagent in the Oppenauer oxidation of alcohols to ketones. It is also used in Meerwein-Ponndorf-Verley reduction of ketones. It is actively involved in the preparation of 1,1,2-triethoxyethene by reacting with 1,1,2,2-tetraethoxyethane followed by the elimination of alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-91-2
Molecular Formula C12H27AlO3-3
Molecular Weight (g/mol) 246.327
MDL Number MFCD00008801
InChI Key BXQSSDQHLWSBEX-UHFFFAOYSA-N
Synonym aluminum tert-butoxide, acmc-20alth, 2-propanol, 2-methyl-,aluminum salt 3:1
PubChem CID 57350944
IUPAC Name aluminum;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].[Al]

Specifications

Melting Point 200°C to 202°C
Density 1.0251 g/mL
Boiling Point 156°C (2 mmHg)
Odor Alcohol-like
Quantity 100 g
UN Number UN2925
Beilstein 4148229
Sensitivity Moisture sensitive
Merck Index 14,333
Solubility Information Very soluble in organic solvents like alcohols,benzene,toluene and xylene.
Formula Weight 246.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Aluminum tert-butoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.