Learn More
Aluminum, SUS, RA19, Ø65x35mm, ARMI∣MBH
Product Code. 30324045
Click to view available options
Quantity:
Ea
Unit Size:
Each
Product Code. 30324045
Brand: ARMI/MBH MBHRA1925
ARMI/MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. Their offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
TRUSTED_SUSTAINABILITY
Specifications
Quantity Ea
Form Disc