missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfide, 99+%, (metals basis)

Product Code. 11347587
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347587 25 g 25g
11357587 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347587 Supplier Thermo Scientific Alfa Aesar Supplier No. 012851.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1302-81-4 | Al2S3 | 150.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1302-81-4
Molecular Formula Al2S3
Molecular Weight (g/mol) 150.14
MDL Number MFCD00014162
InChI Key COOGPNLGKIHLSK-UHFFFAOYSA-N
IUPAC Name dialuminium(3+) trisulfanediide
SMILES [Al+3].[Al+3].[S--].[S--].[S--]

Specifications

Melting Point 1100°C
Color Yellow
Physical Form Pieces
Quantity 25 g
Assay Percent Range >99% (Metal basis)
Formula Weight 150.16
Odor Like rotten eggs
Sensitivity Moisture sensitive
Appearance Yellowish granular
Density 2.02
Chemical Name or Material Aluminum sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.