missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Sulfate Hydrate ACS MP Biomedicals

Product Code. 11430482 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430482 100 g 100g
11440482 500 g 500g
11450482 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430482 Supplier MP Biomedicals Supplier No. 0219140380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17927-65-0
MDL Number MFCD00149136
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
PubChem CID 22377415
ChEBI CHEBI:74779

Specifications

Quantity 100 g
Assay Percent Range 98 to 102%
Molecular Formula Al2O12S3
InChI Key DIZPMCHEQGEION-UHFFFAOYSA-H
SMILES [Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name dialuminium(3+) trisulfate
Molecular Weight (g/mol) 342.13
Formula Weight 342.15 (Anhydrous)
Grade ACS Reagent
Chemical Name or Material Aluminum Sulfate Hydrate, Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.