missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Sulfate Hydrate, ≥97%, Honeywell Fluka™

Product Code. p-7120154 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
100g
2.5kg
500g
Product Code. Quantity Packaging unitSize
15662250 100 g Poly Bottle 100g
15682250 500 g Poly Bottle 500g
15672250 2.5 kg Poly Bottle 2.5kg
3 options
This item is not returnable. View return policy

Product Code. 15662250

Brand: Honeywell-Fluka 227617100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-31-8
Molecular Formula Al2H36O30S3
Molecular Weight (g/mol) 666.401
InChI Key AMVQGJHFDJVOOB-UHFFFAOYSA-H
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
PubChem CID 22377415
ChEBI CHEBI:74779
IUPAC Name dialuminum;trisulfate;octadecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

Melting Point 87°C
Color White
Physical Form Solid
Quantity 100 g
Linear Formula Al2(SO4)3 · 18H2O
UN Number UN3260
Formula Weight 342.15g/mol (anhydrous basis)
Percent Purity ≥97%
Packaging Poly Bottle
Density 1.7g/cm3
Chemical Name or Material Aluminum Sulfate Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.