missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfate hydrate, 97+%, Thermo Scientific Chemicals

Catalog Number 11321749
Click to view available options
Quantity:
500 g
1 kg
5 kg
17927-65-0
Al2O12S3
342.13
MFCD00149136
DIZPMCHEQGEION-UHFFFAOYSA-H
aluminum sulfate hydrate, dialuminum trisulfate hydrate, aluminum sulfate usp, aluminum sulfate 2:3 hydrate, alminum sulfate hydrate, aluminum sulfate usan, sulfuric acid, aluminum salt 3:2 , hydrate, 2al.3so4.h2o
3080712
dialuminium(3+) trisulfate
[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
17927-65-0
Al2O12S3
342.13
MFCD00149136
DIZPMCHEQGEION-UHFFFAOYSA-H
aluminum sulfate hydrate, dialuminum trisulfate hydrate, aluminum sulfate usp, aluminum sulfate 2:3 hydrate, alminum sulfate hydrate, aluminum sulfate usan, sulfuric acid, aluminum salt 3:2 , hydrate, 2al.3so4.h2o
3080712
dialuminium(3+) trisulfate
[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Aluminum sulfate hydrate is a chemical compound commonly used as a flocculating agent in the purification of drinking water and waste water treatment plants, and also in paper manufacturing. Used as coagulant in decolorization and COD removal of secondary yeast wastewater effluents. It is also plays a significant role in arsenic removal by coagulation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Crystalline
Quantity 500 g
Assay Percent Range >97%
Solubility Information It is slightly soluble in alcohol and dilute mineral acids.
Formula Weight 342.15 (Anhydrous)
Odor Odorless
Appearance White crystalline
Density 1.61 g/cm3
Chemical Name or Material Aluminum sulfate hydrate, hydrate
compliance-icons compliance-icons
Product Identifier
  • Aluminum sulfate hydrate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'