missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfate, anhydrous, 99.99% (metals basis)

Product Code. 11475330
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475330 25 g 25g
11465330 5 g 5g
11485330 100 g 100g
11495330 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11475330 Supplier Thermo Scientific Alfa Aesar Supplier No. 044563.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10043-01-3 | Al2O12S3 | 342.131 g/mol

Aluminum sulfate acts as a flocculating agent in the purification of drinking water, and in waste water treatment plants. It acts as a mordant in dyeing, printing, textiles and also used in paper manufacturing. It is a waterproofing agent and accelerator in concrete. Further, it is used as a foaming agent in fire fighting foam, photographic film and in photochemicals. It is also used in styptic pencils, pain relief and in dentistry for gingival retraction cords.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-01-3
Molecular Formula Al2O12S3
Molecular Weight (g/mol) 342.131
MDL Number MFCD00003423
InChI Key DIZPMCHEQGEION-UHFFFAOYSA-H
Synonym aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2
PubChem CID 24850
ChEBI CHEBI:74768
IUPAC Name dialuminum;trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

Melting Point 770°C
pH 3.5
Physical Form Powder
Quantity 25 g
Merck Index 14,366
Solubility Information Soluble in water. Slightly soluble in alcohol and dilute mineral acids. Insoluble in ethanol.
Formula Weight 342.15
Percent Purity 99.99%
Sensitivity Hygroscopic
Density 2.71 g/mL
Assay (metals basis)
Chemical Name or Material Aluminum sulfate, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.