missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Sulfate, 18-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15557844 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
500g
Product Code. Quantity Packaging unitSize
15557844 500 g Poly Bottle 500g
15567844 2.5 kg Poly Bottle 2.5kg
2 options
This item is not returnable. View return policy

Product Code. 15557844

Brand: Avantor J.T.Baker 056401

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7784-31-8
Molecular Formula Al2H36O30S3
Molecular Weight (g/mol) 666.401
InChI Key AMVQGJHFDJVOOB-UHFFFAOYSA-H
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
PubChem CID 22377415
ChEBI CHEBI:74779
IUPAC Name dialuminum;trisulfate;octadecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 666.41
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.69kg
Chemical Name or Material Aluminum Sulfate, 18-Hydrate
compliance-icons
Product Identifier
  • Aluminum sulfate hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contains : Aluminium sulphate,18-hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.