missing translation for 'onlineSavingsMsg'
Learn More

Aluminum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11354388
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
aluminum
[Al]
This item is not returnable. View return policy
7429-90-5
Al
26.98
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
5359268
CHEBI:33629
aluminum
[Al]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 g
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Weight ≈1.1g/slug
Form Slug
Assay (metals basis)
Chemical Name or Material Aluminum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'