missing translation for 'onlineSavingsMsg'
Learn More

Aluminum, silver coated powder, -200 mesh, 99.9% (metals basis), Ag 19-21 wt%, Thermo Scientific Chemicals

Catalog Number 11383908
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
11145-27-0
AlSi
55.067
MFCD05865335
CSDREXVUYHZDNP-UHFFFAOYSA-N
silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
9793685
aluminum;silicon
[Al].[Si]
This item is not returnable. View return policy
11145-27-0
AlSi
55.067
MFCD05865335
CSDREXVUYHZDNP-UHFFFAOYSA-N
silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
9793685
aluminum;silicon
[Al].[Si]

Conductive adhesives

Conductive adhesives

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 25 g
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Ag 19 to 21 wt%
Mesh Size −200 Mesh
Odor Odorless
Weight 1.1g/cm3 to 1.7g/cm3
Form Powder
Assay (metals basis)
Chemical Name or Material Aluminum, Silver coated powder
compliance-icons
Product Identifier
  • Aluminum, silver coated powder
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'