missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aluminum Sign: Evacuation assembly point - ISO 7010

Product Code. 15684197
Click to view available options
Height (Metric):
400 mm
600 mm
Width (Metric):
400 mm
600 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15684197

Brand: Brady™ 138853

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.

  • High performance material for maximum durability
  • Clearly visible and easy to understand

Specifications

Certifications/Compliance ISO 7010
Alternate Part Number E/E007/NT-ALU-400x400/1-B
Material Aluminum
Height (Metric) 400 mm
Thickness 1.016 mm
Width (Metric) 400 mm
Temperature Range (Metric) -40°C to 140°C
Length (Metric) 400 mm
Shape Square
Languages Global
Color White on Green
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.