missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Unmodified HPTLC silica layers on aluminum for TLC. Macherey-Nagel™ Nano-SIL ALUGRAM™ aluminum Sheets feature nano-silica 60 on state-of-the-art aluminum sheets that demonstrate low weight, high strength and temperature stability, and easy cutting.
Unmodified HPTLC silica layers with High Definition properties.
- Binder: highly polymeric product, which is stable in almost all organic solvents and resistant towards aggressive visualization reagents
- Narrow fractionation of the silica particles allows sharper separations, shorter developing times, shorter migration distances, smaller samples and an increased detection sensitivity compared to SIL G plates
- Fluorescent Indicator available: manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
Specifications
Specifications
| Material | Alugram Aluminum Sheets with Nano-SIL Silica Coating |
| Particle Size | 2 to 10 μm |
| Pore Size | 60 Å |
| Quantity | 25 Pk. |
| Type | TLC Plate |
| Fluorescent Indicator | None |
| For Use With (Application) | HPTLC |
| Pore Volume | 0.75 mL/g |
| Specific Surface | ∼500 m2/g |
| Thickness (Metric) | 0.2 mm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?