missing translation for 'onlineSavingsMsg'
Learn More

Aluminum selenide, 99% (metals basis)

Product Code. 11342228
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342228 5 g 5g
11362228 1 g 1g
11352228 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11342228 Supplier Thermo Scientific Alfa Aesar Supplier No. 013132.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1302-82-5

It is used as a precursor to hydrogen selenide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1302-82-5
MDL Number MFCD00014158
Synonym unii-4r7pmp996q, aluminum selenide, aluminum selenide, dialuminum selenium 2-, dialuminium 3+ triselenide, dialuminium 3+ ion triselandiide

Specifications

Color Brown
Physical Form Pieces
Quantity 5 g
UN Number UN3283
Merck Index 14,361
Solubility Information Decomposes to Al203 and SeO2 in water
Formula Weight 290.84
Percent Purity 99%
Odor Odorless
Sensitivity Moisture Sensitive
Density 3.437g/mL
Assay (metals basis)
Chemical Name or Material Aluminum selenide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.