missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sec-butoxide, 95%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog Number 11330318
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
2269-22-9
C12H30AlO3
249.351
MFCD00009327
LWGPRERTOLVRLK-UHFFFAOYSA-N
aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide
50931103
aluminum;butan-2-ol
CCC(C)O.CCC(C)O.CCC(C)O.[Al]
This item is not returnable. View return policy
2269-22-9
C12H30AlO3
249.351
MFCD00009327
LWGPRERTOLVRLK-UHFFFAOYSA-N
aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide
50931103
aluminum;butan-2-ol
CCC(C)O.CCC(C)O.CCC(C)O.[Al]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Density 0.967 g/mL
Boiling Point 200°C to 206°C (30 mmHg)
Flash Point 27°C (82°F)
Odor Alcohol-like
Packaging ChemSeal™ bottles
Refractive Index 1.439
Quantity 100 g
UN Number UN1992
Beilstein 3910908
Sensitivity Moisture sensitive
Formula Weight 246.32
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Aluminum sec-butoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aluminum sec-butoxide
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'