missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sec-butoxide, 95%

Product Code. 11337338
Change view
Click to view available options
Quantity:
10 g
500 g
4 x 500 g
Unit Size:
10g
2000g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337338 4 x 500 g 2000g
11317338 10 g 10g
11347338 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11337338 Supplier Thermo Scientific Alfa Aesar Supplier No. 030721.57
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2269-22-9 | C12H30AlO3 | 249.351 g/mol

It is used for active catalyst and support, inorganic membrane, redrcing agent of aldehyde-ketone, oxidant of aldehyde-ketone, gel forming agent of printing ink and paint, dehydrant, water proofing agent and aluminum base grease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2269-22-9
Molecular Formula C12H30AlO3
Molecular Weight (g/mol) 249.351
MDL Number MFCD00009327
InChI Key LWGPRERTOLVRLK-UHFFFAOYSA-N
Synonym aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide
PubChem CID 50931103
IUPAC Name aluminum;butan-2-ol
SMILES CCC(C)O.CCC(C)O.CCC(C)O.[Al]

Specifications

Density 0.967 g/mL
Boiling Point 200°C to 206°C (30 mmHg)
Flash Point 27°C (82°F)
Odor Alcohol-like
Refractive Index 1.439
Quantity 4 x 500 g
UN Number UN1992
Beilstein 3910908
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water; Soluble in aromatic hydrocarbons, alcohols
Formula Weight 246.32
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Aluminum sec-butoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.