missing translation for 'onlineSavingsMsg'
Learn More

Aluminum rod, 6mm (0.2in) dia, Puratronic™, 99.9965% (metals basis)

Product Code. 11387137
Change view
Click to view available options
Quantity:
15 cm
5 x 15 cm
Unit Size:
15cm
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387137 5 x 15 cm Pack of 5
11397137 15 cm 15cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387137 Supplier Thermo Scientific Alfa Aesar Supplier No. 011478.QY
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx. 0.76g/cm | CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminium
SMILES [Al]

Specifications

Color Silver
Appearance Silver-colored rod
Quantity 5 x 15 cm
Odor Odorless
Assay Percent Range 99.9965% (Metals basis)
Form Rod
Chemical Name or Material Aluminum

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.