missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Reference Material, AA 6061, A96061, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Aluminum, Reference Material, AA 6061, A96061, Ø38x6mm
Specifications
Specifications
| Chemical Name or Material | Zinc/Nickel/Gallium/Vanadium/Titanium/Manganese/Chromium/Copper/Iron/Silicon/Magnesium |
| CAS | 7429-90-5 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Zinc 0.003 %/Nickel 0.01 %/Gallium 0.015 %/Vanadium 0.015 %/Titanium 0.033 %/Manganese 0.076 %/Chromium 0.21 %/Copper 0.29 %/Iron 0.292 %/Silicon 0.619 %/Magnesium 0.878 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction