missing translation for 'onlineSavingsMsg'
Learn More

Kern Sohn Aluminum Protective Box

Product Code. 17391893
Encompass_Preferred
Click to view available options
Classification:
E1 - F2
E1 - M1
E1 - M2
Holds:
1 g
1 g to 1 kg
1 g to 100 g
1 g to 2 kg
1 g to 200 g
1 g to 50 g
1 mg to 10 kg
1 mg to 100 g
1 mg to 2 kg
1 mg to 200 g
1 mg to 5 kg
1 mg to 50 g
1 mg to 500 g
1 mg to 500 mg
2 g
20 kg
50 g
50 kg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17391893

Brand: Kern Sohn 313050600

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Aluminum protected case for standard weight sets.

  • Box for individual weights (cylindrical)
  • For safe storage and transportation under harsh industrial conditions

Specifications

Product Type Case
Classification E1 - M1
Case Material Aluminum
For Use With (Equipment) Button/Compact Weights
Holds 1 mg to 500 g
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.