missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Silicon powder, -325 mesh, 99% (metals basis), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
Description
Aluminum Silicon powder is a reducing agent which is used to make other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Silver to Gray |
Appearance | Silver Gray powder |
Assay Percent Range | 99% (Metals basis) |
Quantity | 100 g |
Solubility Information | Insoluble in water. |
Mesh Size | -325 Mesh |
Odor | Odorless |
Form | Powder |
Chemical Name or Material | Aluminum |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'