missing translation for 'onlineSavingsMsg'
Learn More

Aluminum powder, -100+325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11384658
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384658 50 g 50g
11394658 250 g 250g
11304668 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11384658 Supplier Thermo Scientific Alfa Aesar Supplier No. 043022.18
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifications

Color Silver to Gray
Appearance Silver Gray powder
Quantity 50 g
Ignition Point 400°C (752°F)
Mesh Size -100+325 Mesh
Odor Odorless
Sensitivity Water reactive
Assay Percent Range 99.5% (Metals basis)
Form Powder
IUPAC Name aluminum
Chemical Name or Material Aluminum
Show More Show Less
compliance-icons
Product Identifier
  • Aluminum powder
Signal Word
  • Danger
Hazard Category
  • Pyrophoric solid Category 1
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P223-Do not allow contact with water.
  • P231-Handle under inert gas.
  • P232-Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Aluminium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.