missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate Dodecahydrate, TRC

Product Code. 30136865
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136865 5 g 5g
30132889 25 g 25g
30008068 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136865 Supplier Toronto Research Chemicals Supplier No. TRCA5760485G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlK(SO4)2 . 12H2O
Molecular Weight (g/mol) 474.39
Synonym Alum
IUPAC Name aluminum;potassium;disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/Al.K.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;;2*(H2,1,2,3,4);12*1H2/q+3;+1;;;;;;;;;;;;;;/p-4
Formula Weight 473.9755
Chemical Name or Material Aluminum Potassium Sulfate Dodecahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.