missing translation for 'onlineSavingsMsg'
Learn More

Aluminum potassium sulfate dodecahydrate, ACS, 98.0-102.0%

Product Code. p-4407203
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11373358

Brand: Thermo Scientific Alfa Aesar 036288.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7784-24-9 | AlH24KO20S2 | 474.372 g/mol

In printing, fabrics, dyeing, manufacturing of paper, vegetable glue, baking powder, purifying water, clarifying sugar, as catalyst in ammonia synthesis, hardening agent in microscopyAluminum potassium sulfate dodecahydrate is used in leather tanning, water purification, fire proof textiles and backing powder. In the cosmetic industry, it is used as an after shave treatment, deodorant, hardener for photographic emulsions and as a mordant for dyes. In medical field, it finds application as an astringent, antiseptic and to reduce bleeding in minor cuts and abrasions and nosebleeds..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlH24KO20S2
Molecular Weight (g/mol) 474.372
MDL Number MFCD00149143
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
Synonym kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
PubChem CID 62667
ChEBI CHEBI:86465
IUPAC Name aluminum;potassium;disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Specifications

Melting Point 92°C
Color White
Physical Form Granules
Quantity 500 g
Assay Percent Range 98 -102%
Solubility Information Soluble in water,glycerol. Insoluble in alcoholSoluble in water and glycerol. Insoluble in acetone and alcohol.
Formula Weight 474.38 (258.20 Anhydrous)
Grade ACS
Odor Odorless
Appearance White granular
Density 1.757 g/cm3
Chemical Name or Material Aluminum potassium sulfate dodecahydrate, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.